Print…

Anti-ATP5J Rabbit polyclonal antibody

Fornecedor: Biorbyt
BIRBORB18011-100EA 0 EUR
BIRBORB18011-100
Anti-ATP5J Rabbit polyclonal antibody
Anticorpos
Type: Primary
Antigen: ATP5J (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR