Print…

Anti-BNP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornecedor: Biorbyt
BIRBORB17002-100EA 0 EUR
BIRBORB17002-100
Anti-BNP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: BNP (Brain Natriuretic Peptide)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR