Print…

Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornecedor: Biorbyt
BIRBORB16661-300EA 0 EUR
BIRBORB16661-300
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
Immunofluorescence: 1:50-200

Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR