Print…

Anti-HBsAg Mouse Monoclonal Antibody (FITC (Fluorescein))

Fornecedor: Biorbyt
BIRBORB16520-100EA 0 EUR
BIRBORB16520-100
Anti-HBsAg Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorpos
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: HBsAg
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Virus
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR