Print…

Anti-SMAD4 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornecedor: Biorbyt
BIRBORB16324-100EA 0 EUR
BIRBORB16324-100
Anti-SMAD4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpos
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SMAD4 (SMAD family member 4)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR