Print…

Anti-GEMIN2 Rabbit polyclonal antibody FITC (Fluorescein)

Fornecedor: Biorbyt
BIRBORB16311-100EA 385 EUR
BIRBORB16311-100
Anti-GEMIN2 Rabbit polyclonal antibody FITC (Fluorescein)
Anticorpos
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: GEMIN2 (gem (nuclear organelle) associated protein 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR