Print…

Anti-RRM1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornecedor: Biorbyt
BIRBORB16276-100EA 0 EUR
BIRBORB16276-100
Anti-RRM1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpos
Immunofluorescence: 1:50-200

Type: Primary
Antigen: RRM1 (ribonucleotide reductase M1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR