Print…

Anti-CD163 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB13303-100EA 0 EUR
BIRBORB13303-100
Anti-CD163 Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 500-1000 Immunochemistry: 100-500

Type: Primary
Antigen: CD163 (CD163 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR