|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | ZNF256 |
Antigen name | Zinc Finger Protein 256 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody type | Primary |
Clone | 3H7 |
Host | Mouse |
Isotype | none |
Western blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
Antigen synonyms | BMZF3, BMZF-3 |
Gene ID | 10172 |
Cross adsorption | non |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |