Print…

Anti-DHRS9 Mouse Monoclonal Antibody [clone: 3E3]

Fornecedor: Abnova
ABNOH00010170-M06EA 394 EUR
ABNOH00010170-M06
Anti-DHRS9 Mouse Monoclonal Antibody [clone: 3E3]
Anticorpos
Type: Primary
Antigen: DHRS9
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR