|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | DHRS9 |
Antigen name | Dehydrogenase/reductase (SDR family) Member 9 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Mouse |
Isotype | none |
Western blot | Yes |
Antigen synonyms | RETSDR8, RDHTBE, 3-alpha-HSD, RDHL, RDH15, RDH-E2, RDH-TBE, 3ALPHA-HSD, SDR9C4 |
Gene ID | 10170 |
Cross adsorption | non |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |