Print…

Anti-CHST4 Mouse Polyclonal Antibody

Fornecedor: Abnova
ABNOH00010164-A01EA 258 EUR
ABNOH00010164-A01
Anti-CHST4 Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CHST4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR