Print…

Anti-WASF2 Mouse Monoclonal Antibody [clone: 8E7]

Fornecedor: Abnova
ABNOH00010163-M02EA 394 EUR
ABNOH00010163-M02
Anti-WASF2 Mouse Monoclonal Antibody [clone: 8E7]
Anticorpos
Type: Primary
Antigen: WASF2
Clonality: Monoclonal
Clone: 8E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR