Print…

Anti-FARP1 Mouse Monoclonal Antibody [clone: 2D4]

Fornecedor: Abnova
ABNOH00010160-M01EA 394 EUR
ABNOH00010160-M01
Anti-FARP1 Mouse Monoclonal Antibody [clone: 2D4]
Anticorpos
Type: Primary
Antigen: FARP1
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR