Print…

Anti-TRIM28 Mouse Monoclonal Antibody [clone: 1D11]

Fornecedor: Abnova
ABNOH00010155-M02EA 394 EUR
ABNOH00010155-M02
Anti-TRIM28 Mouse Monoclonal Antibody [clone: 1D11]
Anticorpos
Type: Primary
Antigen: TRIM28
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR