Print…

Anti-ABI2 Rabbit Polyclonal Antibody

Fornecedor: Abnova
ABNOH00010152-D01PEA 380 EUR
ABNOH00010152-D01P
Anti-ABI2 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: ABI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR