Print…

Anti-MBNL2 Mouse Monoclonal Antibody [clone: 3C3]

ABNOH00010150-M01EA 337 EUR
ABNOH00010150-M01
Anti-MBNL2 Mouse Monoclonal Antibody [clone: 3C3]
Type: Primary
Antigen: MBNL2
Clonality: Monoclonal
Clone: 3C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR