Print…
Anti-ARFRP1 Mouse Polyclonal Antibody
Fornecedor: Abnova
ABNOH00010139-B03PEA 394 EUR
ABNOH00010139-B03P
Anti-ARFRP1 Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: ARFRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human