Print…

Anti-ZNF263 Mouse Monoclonal Antibody [clone: 2A1]

Fornecedor: Abnova
ABNOH00010127-M03EA 394 EUR
ABNOH00010127-M03
Anti-ZNF263 Mouse Monoclonal Antibody [clone: 2A1]
Anticorpos
Type: Primary
Antigen: ZNF263
Clonality: Monoclonal
Clone: 2A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR