|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | ACTR1B |
Antigen name | Actin-Related Protein 1B |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody type | Primary |
Clone | 4E10 |
Host | Mouse |
Isotype | none |
ELISA | Yes |
ImmunoFluorescence | Yes |
Antigen synonyms | ARP1B, CTRN2, ARP1 actin-related protein 1 homolog B, centractin beta (yeast), PC3, centractin beta, ARP1, yeast homolog B, ARP1 (actin-related protein 1, yeast) homolog B (centractin beta), ARP1 actin-related protein 1 homolog B, centractin beta |
Gene ID | 10120 |
Cross adsorption | non |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |