Print…
Anti-FEM1B Mouse Polyclonal Antibody
Fornecedor: Abnova
ABNOH00010116-B01EA 380 EUR
ABNOH00010116-B01
Anti-FEM1B Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: FEM1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human