Print…

Anti-TARBP2 Mouse Monoclonal Antibody [clone: 2H1]

Fornecedor: Abnova
ABNOH00006895-M02EA 394 EUR
ABNOH00006895-M02
Anti-TARBP2 Mouse Monoclonal Antibody [clone: 2H1]
Anticorpos
Type: Primary
Antigen: TARBP2
Clonality: Monoclonal
Clone: 2H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR