Print…
Anti-TAF7 Rabbit Polyclonal Antibody
Fornecedor: Abnova
ABNOH00006879-D01PEA 380 EUR
ABNOH00006879-D01P
Anti-TAF7 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: TAF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human