Print…
Anti-TAF7 Mouse Polyclonal Antibody
Fornecedor: Abnova
ABNOH00006879-B01EA 394 EUR
ABNOH00006879-B01
Anti-TAF7 Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: TAF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human