|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | TAF6 |
Antigen name | TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | none |
Western blot | Yes |
Antigen synonyms | MGC:8964, TAFII70, TAF(II)80, TAF2E, TAFII80, TAF(II)70, TAFII-80, TAFII85, TAFII-70 |
Gene ID | 6878 |
Cross adsorption | non |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |