|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | TAGLN |
Antigen name | Transgelin |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | none |
Western blot | Yes |
Antigen synonyms | TAGLN, Transgelin, 22 kDa actin-binding protein, SM22-alpha, Smooth muscle protein 22-alpha, WS3-10, SM22 |
Gene ID | 6876 |
Cross adsorption | non |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |