Print…
Anti-TAF1 Mouse Monoclonal Antibody [clone: 1G9]
Fornecedor: Abnova
ABNOH00006872-M02EA 394 EUR
ABNOH00006872-M02
Anti-TAF1 Mouse Monoclonal Antibody [clone: 1G9]
Anticorpos
Type: Primary
Antigen: TAF1
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human