Print…
Anti-TACR1 Mouse Monoclonal Antibody [clone: 1F7]
Fornecedor: Abnova
ABNOH00006869-M10EA 394 EUR
ABNOH00006869-M10
Anti-TACR1 Mouse Monoclonal Antibody [clone: 1F7]
Anticorpos
Type: Primary
Antigen: TACR1
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human