Print…

Anti-Thr Mouse Monoclonal Antibody [clone: 6D3]

Fornecedor: Abnova
ABNOH00006862-M07EA 394 EUR
ABNOH00006862-M07
Anti-Thr Mouse Monoclonal Antibody [clone: 6D3]
Anticorpos
Type: Primary
Antigen: Thr
Clonality: Monoclonal
Clone: 6D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR