Print…

Anti-Thr Mouse Monoclonal Antibody [clone: 5E11]

Fornecedor: Abnova
ABNOH00006862-M06EA 394 EUR
ABNOH00006862-M06
Anti-Thr Mouse Monoclonal Antibody [clone: 5E11]
Anticorpos
Type: Primary
Antigen: Thr
Clonality: Monoclonal
Clone: 5E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR