Print…

Anti-Thr Rabbit Polyclonal Antibody

Fornecedor: Abnova
ABNOH00006862-D03PEA 380 EUR
ABNOH00006862-D03P
Anti-Thr Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: Thr
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR