Print…
Anti-Thr Mouse Polyclonal Antibody
Fornecedor: Abnova
ABNOH00006862-B01PEA 394 EUR
ABNOH00006862-B01P
Anti-Thr Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: Thr
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human