Print…

Anti-SYN1 Mouse Monoclonal Antibody [clone: 4F6]

Fornecedor: Abnova
ABNOH00006853-M06EA 394 EUR
ABNOH00006853-M06
Anti-SYN1 Mouse Monoclonal Antibody [clone: 4F6]
Anticorpos
Type: Primary
Antigen: SYN1
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR