Print…

Anti-CDH1 Mouse Monoclonal Antibody [clone: OTI2F9]

Fornecedor: OriGene
ORIGTA800691EA 616 EUR
ORIGTA800691
Anti-CDH1 Mouse Monoclonal Antibody [clone: OTI2F9]
Anticorpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR