Print…

Anti-SERBP1 Mouse Monoclonal Antibody [clone: OTI4G2]

Fornecedor: OriGene
ORIGTA800686EA 616 EUR
ORIGTA800686
Anti-SERBP1 Mouse Monoclonal Antibody [clone: OTI4G2]
Anticorpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: SERBP1 (SERPINE1 mRNA binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR