Print…

Anti-CD2 Mouse Monoclonal Antibody [clone: OTI3E11]

Fornecedor: OriGene
ORIGTA800673EA 616 EUR
ORIGTA800673
Anti-CD2 Mouse Monoclonal Antibody [clone: OTI3E11]
Anticorpos
  • Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR