Print…

Anti-MKI67 Mouse Monoclonal Antibody [clone: OTI8H5]

Fornecedor: OriGene
ORIGTA800648EA 616 EUR
ORIGTA800648
Anti-MKI67 Mouse Monoclonal Antibody [clone: OTI8H5]
Anticorpos
Recommended Dilutions: Immunochemistry: 1:50-500; Western Blot: 1:200-1:1000

Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR