|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | ING2 |
Antigen name | Inhibitor of Growth Family, Member 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody type | Primary |
Clone | OTI1E12 |
Host | Mouse |
Isotype | IgG1 |
Western blot | Yes |
Epitope | Full length |
Cross adsorption | none |
Molecular weight | 32.6 kDa |
Purification | purified by affinity chromatography |
Form | Liquid |
Storage buffer | PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide. |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |