Print…

Anti-SF3A1 Mouse Monoclonal Antibody [clone: OTI3F4]

Fornecedor: OriGene
ORIGTA800622EA 616 EUR
ORIGTA800622
Anti-SF3A1 Mouse Monoclonal Antibody [clone: OTI3F4]
Anticorpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: SF3A1 (splicing factor 3a, subunit 1, 120kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR