Print…

Anti-CBX8 Mouse Monoclonal Antibody [clone: OTI3D1]

Fornecedor: OriGene
ORIGTA800617EA 616 EUR
ORIGTA800617
Anti-CBX8 Mouse Monoclonal Antibody [clone: OTI3D1]
Anticorpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: CBX8 (chromobox homolog 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR