Print…

Anti-ZKSCAN4 Mouse Monoclonal Antibody [clone: OTI6C7]

Fornecedor: OriGene
ORIGTA800606EA 616 EUR
ORIGTA800606
Anti-ZKSCAN4 Mouse Monoclonal Antibody [clone: OTI6C7]
Anticorpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZKSCAN4 (zinc finger with KRAB and SCAN domains 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR