Print…

Anti-JUN Mouse Monoclonal Antibody [clone: OTI3C2]

Fornecedor: OriGene
ORIGTA800572EA 616 EUR
ORIGTA800572
Anti-JUN Mouse Monoclonal Antibody [clone: OTI3C2]
Anticorpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR