Print…

Anti-DRD4 Rabbit Polyclonal Antibody

Fornecedor: OriGene
ORIGTA313768EA 543 EUR
ORIGTA313768
Anti-DRD4 Rabbit Polyclonal Antibody
Anticorpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: DRD4 (dopamine receptor D4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR