Print…

Anti-DCC Rabbit Polyclonal Antibody

Fornecedor: OriGene
ORIGTA313747EA 564 EUR
ORIGTA313747
Anti-DCC Rabbit Polyclonal Antibody
Anticorpos
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: DCC (deleted in colorectal carcinoma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR