Print…

Anti-CYP24A1 Rabbit Polyclonal Antibody

Fornecedor: OriGene
ORIGTA313740EA 564 EUR
ORIGTA313740
Anti-CYP24A1 Rabbit Polyclonal Antibody
Anticorpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: CYP24A1 (cytochrome P450, family 24, subfamily A, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR