Print…

Anti-CYP2C18 Rabbit Polyclonal Antibody

Fornecedor: OriGene
ORIGTA313723EA 564 EUR
ORIGTA313723
Anti-CYP2C18 Rabbit Polyclonal Antibody
Anticorpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CYP2C18 (cytochrome P450, family 2, subfamily C, polypeptide 18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR