Print…

Anti-CXADR Rabbit Polyclonal Antibody

Fornecedor: OriGene
ORIGTA313716EA 564 EUR
ORIGTA313716
Anti-CXADR Rabbit Polyclonal Antibody
Anticorpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CXADR (coxsackie virus and adenovirus receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR