Print…

Anti-CSTF1 Rabbit Polyclonal Antibody

Fornecedor: OriGene
ORIGTA313708EA 564 EUR
ORIGTA313708
Anti-CSTF1 Rabbit Polyclonal Antibody
Anticorpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CSTF1 (cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR