Print…

Anti-RASAL1 Goat Polyclonal Antibody

Fornecedor: OriGene
ORIGTA302908EA 550 EUR
ORIGTA302908
Anti-RASAL1 Goat Polyclonal Antibody
Anticorpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: RASAL1 (RAS protein activator like 1 (GAP1 like))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR