Print…

Anti-ZFYVE20 Goat Polyclonal Antibody

Fornecedor: OriGene
ORIGTA302887EA 547 EUR
ORIGTA302887
Anti-ZFYVE20 Goat Polyclonal Antibody
Anticorpos
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-15 µg/ml

Type: Primary
Antigen: ZFYVE20 (zinc finger, FYVE domain containing 20)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR